missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUCLA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576277
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
ATP-specific succinyl-CoA synthetase functions in the citric acid cycle (TCA), coupling the hydrolysis of succinyl-CoA to the synthesis of ATP and thus represents the only step of substrate-level phosphorylation in the TCA. The beta subunit provides nucleotide specificity of the enzyme and binds the substrate succinate, while the binding sites for coenzyme A and phosphate are found in the alpha subunit.
Specifications
| SUCLA2 | |
| Polyclonal | |
| Unconjugated | |
| SUCLA2 | |
| 4930547K18Rik; A-BETA; A-SCS; ATP-specific succinyl-CoA synthetase subunit beta; ATP-specific succinyl-CoA synthetase, beta subunit; mitochondrial succinyl-CoA ligase [ADP-forming] subunit beta; MTDPS5; renal carcinoma antigen NY-REN-39; RP11-528D24.2; SCSbetaA; SCS-betaA; succinate--CoA ligase (ADP-forming); Succinate--CoA ligase [ADP-forming] subunit beta, mitochondrial; succinate-CoA ligase ADP-forming beta subunit; succinate-CoA ligase beta subunit; succinate-CoA ligase, ADP-forming, beta subunit; succinate-Coenzyme A ligase, ADP-forming, beta subunit; succinyl-CoA ligase [ADP-forming] subunit beta, mitochondrial; succinyl-CoA synthetase beta-A chain; Sucla2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20916, 361071, 8803 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P2R7, Q9Z2I9 | |
| SUCLA2 | |
| Recombinant protein of human SUCLA2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction