missing translation for 'onlineSavingsMsg'
Learn More

Succinylcholine chloride, 96%

Product Code. 15505296 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15505296 5 g 5g
15515296 25 g 25g
15525296 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15505296 Supplier Thermo Scientific Acros Supplier No. 460110050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71-27-2 | C14H30Cl2N2O4 | 361.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-27-2
Molecular Formula C14H30Cl2N2O4
Molecular Weight (g/mol) 361.3
InChI Key YOEWQQVKRJEPAE-UHFFFAOYSA-L
Synonym succinylcholine chloride, succinyldicholine chloride, suxamethonium chloride, midarine, scoline, succinylcholine dichloride, anectine, lysthenon, succinylforte, lysthenone
PubChem CID 22475
ChEBI CHEBI:61219
IUPAC Name trimethyl-[2-[4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl]oxyethyl]azanium;dichloride
SMILES C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C.[Cl-].[Cl-]

Specifications

Melting Point 160.0°C to 162.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (on dry substance) (Argentometry)
Quantity 5 g
Solubility Information Solubility in water: soluble.
Formula Weight 361.3
Percent Purity 96%
Chemical Name or Material Succinylcholine chloride
compliance-icons
Product Identifier
  • Succinylcholine chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.