missing translation for 'onlineSavingsMsg'
Learn More

Succinyl sulfathiazole, 99.48%, MP Biomedicals™

Product Code. 11423351 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423351 500 g 500g
11413351 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423351 Supplier MP Biomedicals Supplier No. 0210297690
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Succinylsulfathiazole is used to block the synthesis of folic acid. It is used to study folate deficiency

Chemical Identifiers

CAS 116-43-8
Molecular Formula C13H13N3O5S2
Molecular Weight (g/mol) 355.383
MDL Number MFCD00022437
InChI Key SKVLYVHULOWXTD-UHFFFAOYSA-N
Synonym succinylsulfathiazole, cremosuxidine, sulfasuccidine, sulfasuccinil, sulfasuxidine, colistatin, sulfadigesin, sulfasuccithiazole, sulfasuccidin, sulfenterone
PubChem CID 5315
ChEBI CHEBI:9309
IUPAC Name 4-oxo-4-[4-(1,3-thiazol-2-ylsulfamoyl)anilino]butanoic acid
SMILES C1=CC(=CC=C1NC(=O)CCC(=O)O)S(=O)(=O)NC2=NC=CS2

Specifications

Color White
Quantity 500 g
Formula Weight 355.4
Physical Form Powder
Chemical Name or Material Succinyl sulfathiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.