missing translation for 'onlineSavingsMsg'
Learn More

Succinyl Chloride, ca. 95%

Product Code. p-3737426 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10725971

Brand: Thermo Scientific Acros 132840250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 543-20-4 | C4H4Cl2O2 | 154.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-20-4
Molecular Formula C4H4Cl2O2
Molecular Weight (g/mol) 154.98
MDL Number MFCD00000749
InChI Key IRXBNHGNHKNOJI-UHFFFAOYSA-N
Synonym succinyl chloride, succinyl dichloride, succinic acid dichloride, succinoyl chloride, succinoyl dichloride, succinic chloride, 1,2-bis chlorocarbonyl ethane, butanedioyl chloride, unii-gdn09v9867, 1,2-ethanediylbis chloroformate
PubChem CID 10970
IUPAC Name butanedioyl dichloride
SMILES C(CC(=O)Cl)C(=O)Cl

Specifications

Melting Point 16°C to 17°C
Color Colorless to Brown
Density 1.4070g/mL
Boiling Point 190°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range ca. 95%
Packaging Glass bottle
Linear Formula ClCOCH2CH2COCl
Refractive Index 1.4700 to 1.472
Quantity 25 g
Beilstein 02, IV, 1921
Merck Index 15, 9004
Specific Gravity 1.407
Solubility Information Solubility in water: decomposes
Formula Weight 154.98
Percent Purity ca. 95%
Physical Form Liquid
Chemical Name or Material Succinyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • Succinyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.