missing translation for 'onlineSavingsMsg'
Learn More

Succinic Anhydride, TRC

Product Code. 30011128
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011128 250 g 250g
30008073 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011128 Supplier Toronto Research Chemicals Supplier No. TRCS688770250G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-30-5
Molecular Formula C4 H4 O3
Molecular Weight (g/mol) 100.073
Synonym Dihydro-2,5-furandione, 2,5-Diketotetrahydrofuran, Butanedioic Anhydride, Dihydro-2,5-furandione, NSC 8518, Rikacid SA, Succinic Acid Anhydride, Succinyl Anhydride, Succinyl Oxide, Tetrahydro-2,5-dioxofuran, Tetrahydro-2,5-furandione;
IUPAC Name tetrahydrofuran-2,5-dione
SMILES O=C1CCC(=O)O1

Specifications

Quantity 250 g
InChI Formula InChI=1S/C4H4O3/c5-3-1-2-4(6)7-3/h1-2H2
Formula Weight 100.016
Percent Purity >95%
Chemical Name or Material Succinic Anhydride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.