missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid, MedChemExpress

Product Code. 16459076
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459076 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16459076 Supplier Medchem Express Supplier No. HYN0420
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Succinic acid is a potent and orally active anxiolytic agent. Succinic acid is an intermediate product of the tricarboxylic acid cycle. Succinic acid can be used as a precursor of many industrially important chemicals in food, chemical and pharmaceutical industries.

Succinic acid is a potent and orally active anxiolytic agent. Succinic acid is an intermediate product of the tricarboxylic acid cycle. Succinic acid can be used as a precursor of many industrially important chemicals in food, chemical and pharmaceutical industries.

Specifications

Chemical Name or Material Succinic acid
CAS 110-15-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C4H6O4
Quantity 100 mg
Synonym Wormwood acid
Solubility Information DMSO : 100 mg/mL (846.81 mM; Need ultrasonic) ∣H2O : 30 mg/mL (254.04 mM; Need ultrasonic)
SMILES O=C(O)CCC(O)=O
Molecular Weight (g/mol) 118.09
Formula Weight 118.09
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.