missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinic acid, Free acid, ≥99%, Ultrapure, Thermo Scientific™
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Specifications
Linear Formula | HOOCCH2CH2COOH |
Beilstein | 1754069 |
Formula Weight | 118.09 |
Physical Form | Crystalline Powder |
Density | 1.56 |
Percent Purity | ≥99% |
Boiling Point | 235°C |
Melting Point | 186°C to 188°C |
Quantity | 5 kg |
Flash Point | >110°C |
Show More |
RUO – Research Use Only