missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid, Dr. Ehrenstorfer

Product Code. 30323398
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323398 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323398 Supplier Dr. Ehrenstorfer Supplier No. DREC16985000100MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Succinic acid
CAS 110-15-6
Analyte or Component Names Succinic acid
Shipping Condition +20°C
Molecular Formula C4 H6 O4
Certifications/Compliance ISO 17034
Synonym Succinic Acid, Butanedioic acid
InChI Formula InChI=1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)
SMILES OC(=O)CCC(=O)O
IUPAC Name butanedioic acid
Molecular Weight (g/mol) 118.09
Quantity 100 mg
Formula Weight 118.0266 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.