missing translation for 'onlineSavingsMsg'
Learn More

Succinic Acid Disodium Hexahydrate, MP Biomedicals™

Product Code. 11486332 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486332 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11486332 Supplier MP Biomedicals Supplier No. 0210292991
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use as a TCA (Krebs cycle) supplement in cell culture applications. Succinic Acid Disodium Hexahydrate, MP Biomedicals (aka, sodium succinate dibasic hexahydrate) can be used to study x-ray crystallography, protein structural analysis, proteomics and opti

  • Study x-ray crystallography, protein structural analysis, proteomics and optimization reagents
  • Determine the calorimetric and diffractometric evidence for the sequential crystallization of buffer components and the consequential pH swing in frozen solutions
  • Provide insights into the mechanism of Pseudomonas dacunhae aspartate β-decarboxylase from rapid-scanning stopped-flow kinetics

Specifications

CAS 6106-21-4
Quantity 1 kg
Physical Form Crystalline Powder
Molecular Formula C4H16Na2O10
Synonym disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
Solubility Information Soluble in water (100mg/mL, clear and colorless solution).
InChI Key ZBTUYCUNQBRXOR-UHFFFAOYSA-L
SMILES C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]
IUPAC Name disodium;butanedioate;hexahydrate
Molecular Weight (g/mol) 270.142
PubChem CID 3083938
ChEBI CHEBI:63686
Formula Weight 270.1
Percent Purity ≥98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.