missing translation for 'onlineSavingsMsg'
Learn More

Succinic Acid (Butanedioic Acid), Mikromol™

Product Code. 30051697
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051697 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051697 Supplier Mikromol Supplier No. MM0567.050100
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Succinic Acid (Butanedioic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Succinic acid
CAS 110-15-6
Analyte or Component Names Succinic acid
Shipping Condition Room Temperature
Molecular Formula C4 H6 O4
Certifications/Compliance ISO 9001
Synonym Succinic Acid, Butanedioic acid
InChI Formula InChI=1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)
SMILES OC(=O)CCC(=O)O
IUPAC Name butanedioic acid
Molecular Weight (g/mol) 118.09
Quantity 100 mg
Formula Weight 118.0266 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.