missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid, ACS, ≥99%, MP Biomedicals™

Product Code. 11404682 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404682 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11404682 Supplier MP Biomedicals Supplier No. 0219144980
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 110-15-6
Color White
Quantity 100 g
Physical Form Powder
Assay Percent Range ≥99%
Molecular Formula C4H6O4
MDL Number MFCD00002789
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
SMILES OC(=O)CCC(O)=O
IUPAC Name butanedioic acid
Molecular Weight (g/mol) 118.09
PubChem CID 1110
ChEBI CHEBI:15741
Formula Weight 118.09
Chemical Name or Material Succinic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.