missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid, 99+%

Product Code. 11400003
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400003 500 g 500g
11410003 2500 g 2500g
11420003 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400003 Supplier Thermo Scientific Alfa Aesar Supplier No. A12084.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-15-6 | C4H6O4 | 118.09 g/mol

Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry, as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry, as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.

Solubility
Soluble in ethanol, ethyl ether, acetone and methanol. Insoluble in toluene, benzene, carbon disulfide, carbon tetrachloride and petroleum ether.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
Synonym succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid
PubChem CID 1110
ChEBI CHEBI:15741
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

Melting Point 186°C to 188°C
Boiling Point 235°C
Beilstein 1754069
Merck Index 14,8869
Solubility Information Soluble in ethanol,ethyl ether,acetone and methanol. Insoluble in toluene,benzene,carbon disulfide,carbon tetrachloride and petroleum ether.
Formula Weight 118.09
Percent Purity ≥99%
Density 1.56
Quantity 500 g
Flash Point 206°C (402°F)
Chemical Name or Material Succinic acid
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Butanedioic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.