missing translation for 'onlineSavingsMsg'
Learn More

Succinamic acid, 97%

Product Code. 10562941 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10562941 100 g Glass bottle 100g
10552941 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10562941 Supplier Thermo Scientific Acros Supplier No. 158731000
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 638-32-4 | C4H7NO3 | 117.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-32-4
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.10
MDL Number MFCD00008041
InChI Key JDVPQXZIJDEHAN-UHFFFAOYSA-N
Synonym succinamic acid, butanoic acid, 4-amino-4-oxo, succinic acid monoamide, 3-carbamoylpropanoic acid, succinamidic acid, acmc-1b64y, 3-c-hydroxycarbonimidoyl propanoic acid
PubChem CID 12522
ChEBI CHEBI:50398
IUPAC Name 4-amino-4-oxobutanoic acid
SMILES NC(=O)CCC(O)=O

Specifications

Melting Point 153°C to 157°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2NCOCH2CH2CO2H
Quantity 100 g
Beilstein 02, 614
Formula Weight 117.1
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material Succinamic acid, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.