missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Substance P Antibody (JE59-73), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32984
This item is not returnable.
View return policy
Description
Substance P Monoclonal antibody specifically detects Substance P in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Substance P | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 | |
| neurokinin 1, neurokinin 2, neurokinin A, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma), neuropeptide gamma, neuropeptide K, NK2, NKA, NKNAneurokinin alpha, NPK, PPT, protachykinin-1, substance K, substance P, TAC2neuromedin L, tachykinin 2, tachykinin, precursor 1, tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2 | |
| Synthetic peptide within Human Substance P aa 41-90/ 129. (Uniprot: P20366) | |
| 100 μL | |
| Growth and Development, Neuroendocrinology, Neuroscience | |
| 6863 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| JE59-73 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction