Learn More
Substance P, Thermo Scientific Chemicals
For cellular and molecular biology applications
Brand:  Thermo Scientific Alfa Aesar J63489.MB
Code : NEW
Additional Details : CAS Number : 33507-63-0 Weight : 0.00025kg
 View more versions of this product
Product Code. 15405719
  • £557.00 / 25mg
Estimated Shipment: 29-05-2024
to see stock.
Description
Description
Substance P is used as a neurokinin NK-1 receptor agonist, neurokinin undecapeptide neurotransmitter and neuromodulator, also used in cancer cell proliferation and that inhibition of this component can induce apoptosis in cancer cells and decrease the steady state of Her2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
HPLC
A neurokinin NK-1 receptor agonist
Substance P
RUO - research use only
Powder
1347.64
25 mg
Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2
Product Suggestions
Product Suggestions
Videos
SDS Documents