missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ SUBHT High Throughput Submarine Gel Unit
Click to view available options
Type:
Buffer Chamber Assembly
Casting Gate
Colored Loading Strip
High Throughput Submarine Gel Unit
Labeled Running Tray
Lid
Replacement High Voltage Lead
Running Tray
Silicone Gasket
Unit Size:
Each
Description
Includes
- Buffer chamber assembly
- Safety lid, high-voltage leads
- Fluorescent Labeled Running Tray with Casting Gates
- Four 1mm thick 30-well combs
- Colored Loading Strips
- Two Buffer Recirculation Ports
- Fast, error-free loading: load an entire 96-well plate using a multichannel pipetter
- Simplified Analysis: Fluorescent labels for each sample printed on a UV transparent gel tray allows for instant sample identification (matched to a 96-well plate) during post-run visualization
- High Throughput: Run up to 120 samples (96-well plate plus 24 control and/or marker samples) in 30 minutes
- Better Reproducibility: Eliminate variability caused by running buffer pH drift and ionic gradient formation with optional buffer recirculation capability
- Reduced Buffer Needs: Compact tank design needs less buffer to cover gel which not only reduces buffer consumption but provides greater control of voltage gradients and run times
- Maximum Flexibility: Fully compatible with a wide range of combs and accessories to meet all your high throughput and standard gel analysis needs
- This high throughput horizontal gel unit can accommodate up to 120 samples and resolve DNA strands from 50-1000 base pairs in 30 minutes
- Gel Dimensions: 15 x 15 cm
- Maximum Buffer Volume: 1.2L
- Maximum Power Settings: 300V, 200mA, 60W
- Maximum Sample Capacity: 120
- Maximum Running Temperature: 45°C
- Indoor Use: 4-40°C
- Humidity: up to 80%
- Unit Dimensions: 33.5L x 21.5W x 7 cmH
- Safety Certifications: EN61010-1, CE
Applications:
Single Strand Polymorphism (SSP) analysis
Specifications
Specifications
| For Use With (Equipment) | SUBHT High Throughput Submarine Gel Unit |
| Type | Colored Loading Strip |
| Includes | Buffer chamber assembly, safety lid with high voltage Leads, fluorescent labeled running tray with casting gates, four 1mm thick 30-well combs, colored loading strips, two Buffer recirculation ports |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction