missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ SUBHT High Throughput Submarine Gel Unit

Product Code. 12625826
Change view
Click to view available options
Type:
Buffer Chamber Assembly
Casting Gate
Colored Loading Strip
High Throughput Submarine Gel Unit
Labeled Running Tray
Lid
Replacement High Voltage Lead
Running Tray
Silicone Gasket
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
12625826 Running Tray Each
12675816 Colored Loading Strip Each
12655816 High Throughput Submarine Gel Unit Each
12615826 Buffer Chamber Assembly Each
12655826 Silicone Gasket Each
12665816 Casting Gate Each
12635826 Replacement High Voltage Lead Each
12695816 Lid Each
12605826 Labeled Running Tray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 12625826 Supplier Hoefer™ Supplier No. SUBHTUT
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

  • Buffer chamber assembly
  • Safety lid, high-voltage leads
  • Fluorescent Labeled Running Tray with Casting Gates
  • Four 1mm thick 30-well combs
  • Colored Loading Strips
  • Two Buffer Recirculation Ports

Ideal for high throughput screening applications like single strand polymorphism (SSP) analysis following PCR

  • Fast, error-free loading: load an entire 96-well plate using a multichannel pipetter
  • Simplified Analysis: Fluorescent labels for each sample printed on a UV transparent gel tray allows for instant sample identification (matched to a 96-well plate) during post-run visualization
  • High Throughput: Run up to 120 samples (96-well plate plus 24 control and/or marker samples) in 30 minutes
  • Better Reproducibility: Eliminate variability caused by running buffer pH drift and ionic gradient formation with optional buffer recirculation capability
  • Reduced Buffer Needs: Compact tank design needs less buffer to cover gel which not only reduces buffer consumption but provides greater control of voltage gradients and run times
  • Maximum Flexibility: Fully compatible with a wide range of combs and accessories to meet all your high throughput and standard gel analysis needs
  • This high throughput horizontal gel unit can accommodate up to 120 samples and resolve DNA strands from 50-1000 base pairs in 30 minutes
Specifications
  • Gel Dimensions: 15 x 15 cm
  • Maximum Buffer Volume: 1.2L
  • Maximum Power Settings: 300V, 200mA, 60W
  • Maximum Sample Capacity: 120
  • Maximum Running Temperature: 45°C
  • Indoor Use: 4-40°C
  • Humidity: up to 80%
  • Unit Dimensions: 33.5L x 21.5W x 7 cmH
  • Safety Certifications: EN61010-1, CE

Applications:

Single Strand Polymorphism (SSP) analysis

Specifications

For Use With (Equipment) SUBHT High Throughput Submarine Gel Unit
Type Running Tray
Includes Casting gate
Dimensions (L x W) 5.9 x 5.9 in. (15 x 15 cm)
Length (English) 5.9 in.
Length (Metric) 15 cm
Width (English) 5.9 in.
Width (Metric) 15 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.