missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Subframe on Castors for UN75plus
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN75plus Oven
Unit Size:
Each
Description
- On Castors
- Height: 660 mm
Specifications
Specifications
| Description | On Wheels |
| For Use With (Equipment) | UN75plus Oven |
| Type | Subframe |
| Height (Metric) | 660 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction