Learn More
Memmert™ Subframe on Castors for UF160plus
Product Code. 17559055 Shop All Memmert Products
Click to view available options
Description:
On Wheels
Unit Size:
Each
Product Code. 17559055
Brand: Memmert™ UF160plusB29750
Only null left
Add to basket
Add to basket
Subframe on wheels for Memmert Ovens.
  • On Castors
  • Height: 560 mm
Specifications
Description On Wheels
For Use With (Equipment) UF160plus Oven
Type Subframe
Height (Metric) 560 mm