missing translation for 'onlineSavingsMsg'
Learn More

Suberic Acid, TRC

Product Code. 30134163
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134163 10 g 10g
17914895 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134163 Supplier Toronto Research Chemicals Supplier No. TRCS68867010G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-48-6
Molecular Formula C8 H14 O4
Molecular Weight (g/mol) 174.19
Synonym Octanedioic acid (6CI, 9CI, ACI), Suberic acid (8CI), 1,6-Dicarboxyhexane, 1,6-Hexanedicarboxylic acid, 1,8-Octanedioic acid, Cork acid, Hexamethylenedicarboxylic acid, NSC 25952, NSC 53777
IUPAC Name octanedioic acid
SMILES OC(=O)CCCCCCC(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H14O4/c9-7(10)5-3-1-2-4-6-8(11)12/h1-6H2,(H,9,10)(H,11,12)
Formula Weight 174.0892
Chemical Name or Material Suberic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.