Learn More
Suberic acid, 99%
Description
Suberic acid is used in drug synthesis and in the production of plastics. It is also used in the preparation of reduction-sensitive micelles, which affects their cellular uptake. Further, it is used in the delivery of anticancer drugs. In addition, it is used in the fluorescent detection of amidinium-carboxylate and amidinium formation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suberic acid is used in drug synthesis and in the production of plastics. It is also used in the preparation of reduction-sensitive micelles, which affects their cellular uptake. Further, it is used in the delivery of anticancer drugs. In addition, it is used in the fluorescent detection of amidinium-carboxylate and amidinium formation.
Solubility
Soluble in water,dimethyl sulfoxide and methanol. Slightly soluble in ether. Insoluble in chloroform.
Notes
Incompatible with strong oxidizing agents, reducing agents and bases.
Specifications
Specifications
| Melting Point | 141°C to 144°C |
| Boiling Point | 230°C (15 mmHg) |
| Flash Point | 203°C (397°F) |
| Quantity | 2500 g |
| Beilstein | 1210161 |
| Merck Index | 14,8862 |
| Solubility Information | Soluble in water,dimethyl sulfoxide and methanol. Slightly soluble in ether. Insoluble in chloroform. |
| Formula Weight | 174.2 |
| Percent Purity | 99% |
| Chemical Name or Material | Suberic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.