missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10948-2-AP-20UL
This item is not returnable.
View return policy
Description
This antibody recognize 14-16 kDa (P14 or P15), 20 kDa (phospho form) and 30 kDa (Dimer form).
PC4 (activated RNA polymerase II transcription cofactor 4) is a transcriptional coactivator, possessing the ability to suppress promoter-driven as well as nonspecific transcription via its DNA binding activity. The repressive activity of PC4 on promoter-driven transcription is alleviated by transcription factor TFIIH. TFIIH protects promoters from PC4-mediated repression by relieving the topological constraint imposed by PC4 through the ERCC3 helicase activity.Specifications
| SUB1 | |
| Polyclonal | |
| Unconjugated | |
| SUB1 | |
| p14, PC4, Positive cofactor 4, RPO2TC1, SUB1, SUB1 homolog, SUB1 homolog (S. cerevisiae) | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10923, 192269, 20024 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P11031, P53999, Q63396 | |
| Sub1 | |
| SUB1 Fusion Protein Ag1390 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction