missing translation for 'onlineSavingsMsg'
Learn More

SUB1 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18355755 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355755 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355755 Supplier Novus Biologicals Supplier No. NBP300275DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SUB1 Polyclonal specifically detects SUB1 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUB1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias activated RNA polymerase II transcription cofactor 4, activated RNA polymerase II transcriptional coactivator p15, MGC102747, p15, PC4p14P15, Positive cofactor 4, RPO2TC1, SUB1 homolog, SUB1 homolog (S. cerevisiae)
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant SUB1 (P53999; Met1-Leu127).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 10923
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.