missing translation for 'onlineSavingsMsg'
Learn More

SUB1 Antibody (3A1), Novus Biologicals™

Product Code. 18327139 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327139 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327139 Supplier Novus Biologicals Supplier No. H00010923M11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SUB1 Monoclonal antibody specifically detects SUB1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUB1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3A1
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH09610
Gene Alias activated RNA polymerase II transcription cofactor 4, activated RNA polymerase II transcriptional coactivator p15, MGC102747, p15, PC4p14P15, Positive cofactor 4, RPO2TC1, SUB1 homolog, SUB1 homolog (S. cerevisiae)
Host Species Mouse
Immunogen SUB1 (AAH09610.1, 32 a.a. ~ 124 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. APEKPVKKQKTGETSRALSSSKQSSSSRDDNMFQIGKMRYVSVRDFKGKVLIDIREYWMDPEGEMKPGRKGISLNPEQWSQLKEQISDIDDAV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 10923
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.