missing translation for 'onlineSavingsMsg'
Learn More

SU1498, MedChemExpress

Product Code. 16487728
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487728 10 mg 10mg
16461799 1 mg 1mg
16433707 50 mg 50mg
16433636 25 mg 25mg
16410198 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16487728 Supplier Medchem Express Supplier No. HY19326
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SU1498 (AG 1498) is a selective inhibitor of the VEGFR2; inhibits Flk-1 with an IC50 of value of 700 nM.

SU1498 (AG 1498) is a selective inhibitor of the VEGFR2

Specifications

Chemical Name or Material SU1498
CAS 168835-82-3
Color White
Purity Grade Notes Research
Molecular Formula C25H30N2O2
Quantity 10 mg
Synonym AG 1498 Tyrphostin SU 1498
Solubility Information DMSO : 100 mg/mL (256.07 mM; Need ultrasonic)
SMILES O=C(NCCCC1=CC=CC=C1)/C(C#N)=C/C2=CC(C(C)C)=C(O)C(C(C)C)=C2
Molecular Weight (g/mol) 390.52
Formula Weight 390.52
Percent Purity 98.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.