missing translation for 'onlineSavingsMsg'
Learn More

SU11274, MedChemExpress

Product Code. 16465696
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465696 100 mg 100mg
16444918 10 mg 10mg
16441167 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16465696 Supplier Medchem Express Supplier No. HY12014
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SU11274 is a selective Met inhibitor with IC50 of 10 nM, but has no effects on PGDFRβ, EGFR or Tie2.

SU11274 is a selective Met inhibitor with IC50 of 10 nM, but has no effects on PGDFRβ, EGFR or Tie2.

Specifications

Chemical Name or Material SU11274
CAS 658084-23-2
Color Orange
Purity Grade Notes Research
Molecular Formula C28H30ClN5O4S
Quantity 100 mg
Synonym PKI-SU11274
Solubility Information DMSO : ≥ 100 mg/mL (176.03 mM)
SMILES O=S(C1=CC2=C(C=C1)NC(/C2=C/C3=C(C(C(N4CCN(CC4)C)=O)=C(N3)C)C)=O)(N(C)C5=CC=CC(Cl)=C5)=O
Molecular Weight (g/mol) 568.09
Formula Weight 568.09
Percent Purity 98.19%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.