missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STYXL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562159
This item is not returnable.
View return policy
Description
Immunogen sequence: PGLLLCEPTE LYNILNQATK LSRLTDPNYL CLLDVRSKWE YDESHVITAL RVKKKNNEYL LPESVDLECV KYCVVYDNNS STLEILLKDD DDDSDSDGDG KD Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.
Catalytically inactive phosphatase (PubMed:20180778, PubMed:23163895). By binding to G3BP1, inhibits the formation of G3BP1-induced stress granules (PubMed:20180778, PubMed:23163895). Does not act by protecting the dephosphorylation of G3BP1 at 'Ser-149' (PubMed:23163895). Inhibits PTPMT1 phosphatase activity (PubMed:24709986). By inhibiting PTPMT1, positively regulates intrinsic apoptosis (PubMed:21262771). May play a role in the formation of neurites during neuronal development (PubMed:29250526). [UniProt].
Specifications
| STYXL1 | |
| Polyclonal | |
| Unconjugated | |
| STYXL1 | |
| dual specificity phosphatase 24 (putative); dual specificity phosphatase inhibitor MK-STYX; Dual specificity protein phosphatase 24; DUSP24; Inactive dual specificity protein phosphatase MK-STYX; Map kinase phosphatase-like protein MK-STYX; MKSTYX; MK-STYX; serine/threonine/tyrosine interacting-like 1; serine/threonine/tyrosine-interacting-like protein 1; STYXL1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51657 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y6J8 | |
| STYXL1 | |
| Recombinant protein corresponding to Human STYXL1. Recombinant protein control fragment (Product #RP-91559). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction