missing translation for 'onlineSavingsMsg'
Learn More

Styrene (stabilized with 4-tert-Butylcatechol), TRC

Product Code. 30137322
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137322 5 g 5g
30138583 25 g 25g
30040834 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137322 Supplier Toronto Research Chemicals Supplier No. TRCS6877905G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-42-5
Molecular Formula C8 H8
Molecular Weight (g/mol) 104.15
Synonym Styrene, Benzene, ethenyl-, Styrene (8CI), Ethenylbenzene, BYX 6, Cinnamene, EM 50, Maomin SM, NSC 62785, Phenethylene, Phenylethene, Phenylethylene, S 0095, STC, Styrol, Styrole, Styrolene, Styrolution PS 124N, Styropol SO, TTB 7302, Vinylbenzene, Vinylbenzol
IUPAC Name styrene
SMILES C=Cc1ccccc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2
Formula Weight 104.0626
Percent Purity >95%
Chemical Name or Material Styrene
Grade HPLC
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.