missing translation for 'onlineSavingsMsg'
Learn More

Styrene-d{8}, 98% (Isotopic) stab. with 4-tert-butylcatechol

Product Code. 11395887
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395887 1 g 1g
11305897 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11395887 Supplier Thermo Scientific Alfa Aesar Supplier No. 043667.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 19361-62-7 | C8H8 | 112.201 g/mol

Styrene-d{8} is used in Organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19361-62-7
Molecular Formula C8H8
Molecular Weight (g/mol) 112.201
MDL Number MFCD00044231
InChI Key PPBRXRYQALVLMV-GDALLCCDSA-N
Synonym styrene-d8, 2h8 styrene, perdeuteriostyrene, benzene-d5, ethenyl-d3, 1,2,3,4,5-pentadeuterio-6-1,2,2-trideuterioethenyl benzene, benzene-1,2,3,4,5-d5, 6-ethenyl-1,2,2-d3, styrene d8, phenethylene-d8, phenylethene-d8, vinylbenzene-d8
PubChem CID 88025
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(1,2,2-trideuterioethenyl)benzene
SMILES C=CC1=CC=CC=C1

Specifications

Physical Form Liquid
Color Colorless
Density 0.979 g/mL
Flash Point 31°C (88°F)
Packaging Ampule
Refractive Index 1.5445
Sensitivity Air
Quantity 1 g
Boiling Point 50°C to 51°C (25mmHg)
Linear Formula C6D5CD=CD2
UN Number UN2055
Beilstein 2327045
Solubility Information It is soluble in water 0.24g/L (20°C).
Formula Weight 112.2
Percent Purity 98%
Chemical Name or Material Styrene-d8, Stabilized with 4-tert-butylcatechol, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.