missing translation for 'onlineSavingsMsg'
Learn More

Styrene-4-sulfonic acid sodium salt hydrate, tech. 90%

Product Code. 11382567
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382567 25 g 25g
11372567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382567 Supplier Thermo Scientific Alfa Aesar Supplier No. L11685.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2695-37-6 | C8H7NaO3S | 206.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2695-37-6
Molecular Formula C8H7NaO3S
Molecular Weight (g/mol) 206.19
MDL Number MFCD00013379 MFCD00084449
InChI Key XFTALRAZSCGSKN-UHFFFAOYSA-M
Synonym sodium 4-vinylbenzenesulfonate, sodium p-styrenesulfonate, sodium 4-styrenesulfonate, sodium p-styrene sulfonate, unii-0kp0v3og5g, benzenesulfonic acid, 4-ethenyl-, sodium salt, poly sodium 4-vinylbenzenesulfonate, 0kp0v3og5g, sodium 4-ethenylbenzenesulfonate, p-styrenesulfonic acid sodium salt
PubChem CID 3571582
IUPAC Name sodium;4-ethenylbenzenesulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=C(C=C)C=C1

Specifications

Melting Point >320°C
Quantity 25 g
Beilstein 3575931
Formula Weight 206.20 (Anhydrous)
Percent Purity 90%
Grade Technical
Chemical Name or Material Styrene-4-sulfonic acid sodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.