missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Styrene-4-sulfonic acid sodium salt, Thermo Scientific Chemicals
99.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
Description
Styrene-4-sulfonic acid sodium salt is grafted on poly(ethersulfone) membrane to determine the fractionation of proteins with similar molecular weights. It is used as sodium based ion exchange membranes for membrane capacitive deionization (MCDI) for synthesis. It is used in the study of copolymerization characteristics of sodium styrenesulfonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Quantity | 250 g |
Beilstein | 3575931 |
Solubility Information | Soluble in water. |
Formula Weight | 206.19 |
Physical Form | Powder |
Chemical Name or Material | Styrene-4-sulfonic acid sodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'