missing translation for 'onlineSavingsMsg'
Learn More

STYK1 Antibody (3A8), Novus Biologicals™

Product Code. 18334679 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334679 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334679 Supplier Novus Biologicals Supplier No. H00055359M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

STYK1 Monoclonal antibody specifically detects STYK1 in Human samples. It is validated for Western Blot, ELISA, ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen STYK1
Applications Western Blot, ELISA, Sandwich ELISA, KnockDown
Classification Monoclonal
Clone 3A8
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_060893
Gene Alias DKFZp761P1010, EC 2.7.10.2, NOKProtein PK-unique, serine/threonine/tyrosine kinase 1Novel oncogene with kinase domain, SuRTK106, tyrosine-protein kinase STYK1
Host Species Mouse
Immunogen STYK1 (NP_060893, 50 a.a. ~ 159 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. REQRTQQQRSGPQGIAPVPPPRDLSWEAGHGGNVALPLKETSVENFLGATTPALAKLQVPREQLSEVLEQICSGSCGPIFRANMNTGDPSKPKSVILKALKEPAGLHEVQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 55359
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.