Learn More
STXBP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15312-1-AP-20UL
 View more versions of this product
Product Code. 16814444
  • £135.00 / 20µL
Description
This gene encodes a member of the STXBP/unc-18/SEC1 family. The encoded protein is involved in intracellular trafficking, control of SNARE (soluble NSF attachment protein receptor) complex assembly, and the release of cytotoxic granules by natural killer cells. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Specifications
Specifications
STXBP2
Polyclonal
Unconjugated
STXBP2
Hunc18b, MUNC18 2, pp10122, STXBP2, STXBP2 protein, syntaxin binding protein 2, UNC18 2, UNC18B
Rabbit
Antigen Affinity Chromatography
RUO
20911, 6813
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15833, Q64324
STXBP2
STXBP2 Fusion Protein Ag7537
20 μL
Primary
Mouse, Human