Learn More
STX5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26711-1-AP-150UL
This item is not returnable.
View return policy
Description
The STX5 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human STX5. This antibody recognizes human, mouse antigen. The STX5 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| STX5 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 6811 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 267 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |