missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STX5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5112287
This item is not returnable.
View return policy
Description
Mediates endoplasmic reticulum to Golgi transport. Together with p115/USO1 and GM130/GOLGA2, involved in vesicle tethering and fusion at the cis-Golgi membrane to maintain the stacked and inter-connected structure of the Golgi apparatus.
Specifications
| STX5 | |
| Polyclonal | |
| Unconjugated | |
| STX5 | |
| 0610031F24Rik; D19Ertd627e; SED5; Stx5; Stx5a; syntaxin 5; syntaxin 5A; syntaxin-5 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 6811 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q13190 | |
| STX5 | |
| E. coli-derived Human Stx5 fragment | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction