Learn More
Invitrogen™ STX18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551797
Description
Immunogen sequence: VKRVVDKKRL SKLEPEPNTK TRESTSSEKV SQSPSKDSEE NPATEERPEK ILAETQPELG TWGDGKGEDE LSPEEIQMFE QENQRLIGEM NSLFDEVRQI EGRVVEISRL QEIFTEKVLQ QEAEIDSIHQ LVVGATENIK EGNEDIREAI Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
Syntaxins, a family of proteins involved in the fusion of synaptic vesicles with the plasma membrane, display broad tissue distribution and contain carboxy-terminal hydrophobic domains that direct themselves to their respective intracellular compartments. Syntaxin 18 mainly localizes to the endoplasmic reticulum (ER), and functions in transport between the ER and Golgi. Syntaxin 18 also plays a role in ER-mediated phagocytosis, possibly by regulating the specific fusion of the ER and plasma or phagosomal membranes.
Specifications
| STX18 | |
| Polyclonal | |
| Unconjugated | |
| STX18 | |
| 1810035L21Rik; 4933425D03Rik; AU041562; cell growth-inhibiting gene 9 protein; GIG9; STX18; syntaxin 18; syntaxin-18; Ufe1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 53407 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P2W9 | |
| STX18 | |
| Recombinant protein corresponding to Human STX18. Recombinant protein control fragment (Product #RP-95933). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.