missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STX18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550799
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total STX18 protein.
Syntaxins, a family of proteins involved in the fusion of synaptic vesicles with the plasma membrane, display broad tissue distribution and contain carboxy-terminal hydrophobic domains that direct themselves to their respective intracellular compartments. Syntaxin 18 mainly localizes to the endoplasmic reticulum (ER), and functions in transport between the ER and Golgi. Syntaxin 18 also plays a role in ER-mediated phagocytosis, possibly by regulating the specific fusion of the ER and plasma or phagosomal membranes.
Specifications
| STX18 | |
| Polyclonal | |
| Unconjugated | |
| STX18 | |
| 1810035L21Rik; 4933425D03Rik; AU041562; cell growth-inhibiting gene 9 protein; GIG9; STX18; syntaxin 18; syntaxin-18; Ufe1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53407, 71116 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.4 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8VDS8, Q9P2W9 | |
| STX18 | |
| Full length fusion protein of Human STX18. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction