missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STX18 Monoclonal Antibody (OTI2C4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527135
This item is not returnable.
View return policy
Description
STX18 Monoclonal Antibody for Western Blot
Syntaxins, a family of proteins involved in the fusion of synaptic vesicles with the plasma membrane, display broad tissue distribution and contain carboxy-terminal hydrophobic domains that direct themselves to their respective intracellular compartments. Syntaxin 18 mainly localizes to the endoplasmic reticulum (ER), and functions in transport between the ER and Golgi. Syntaxin 18 also plays a role in ER-mediated phagocytosis, possibly by regulating the specific fusion of the ER and plasma or phagosomal membranes.
Specifications
| STX18 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9P2W9 | |
| STX18 | |
| Human recombinant protein fragment corresponding to amino acids 1-285 of STX18 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2C4 | |
| Unconjugated | |
| STX18 | |
| 1810035L21Rik; 4933425D03Rik; AU041562; cell growth-inhibiting gene 9 protein; GIG9; STX18; syntaxin 18; syntaxin-18; Ufe1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 53407 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction