missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STX16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559395
This item is not returnable.
View return policy
Description
Immunogen sequence: SPLHSRSIAA ELDELADDRM ALVSGISLDP EAAIGVTKRP PPKWVDGVDE IQYDVGRIKQ KMKELASLHD KHLNRPTLDD SSEEEHAIEI TTQEITQLFH R Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 87%.
This gene encodes a protein that is a member of the syntaxin or t-SNARE (target-SNAP receptor) family. These proteins are found on cell membranes and serve as the targets for V-SNARES (vesicle-SNAP receptors) permitting specific synaptic vesicle docking and fusion. A microdeletion in the region of chromosome 20 where this gene is located has been associated with pseudohypoparathyroidism type Ib. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| STX16 | |
| Polyclonal | |
| Unconjugated | |
| STX16 | |
| 4930401D03; 5430410K23Rik; 6330500A18Rik; AI648908; AW553605; CG1467; CG1467-PA; Dm Syx16; Dmel\CG1467; Dmel_CG1467; DmSyx16; dSyntaxin16; dSyx 16; dSyx16; MGC90328; RP11-261P9.3; STX16; Syn16; syntaxin; syntaxin 16; Syntaxin16; syntaxin-16; Syx; SYX16; Syx16-PA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8675 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14662 | |
| STX16 | |
| Recombinant protein corresponding to Human STX16. Recombinant protein control fragment (Product #RP-98449). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido