missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STX16 Polyclonal Antibody
GREENER_CHOICE

Sheep Polyclonal Antibody

Brand:  Invitrogen™ PA5149042

Product Code. 30534661

  • £415.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

This gene encodes a protein that is a member of the syntaxin or t-SNARE (target-SNAP receptor) family. These proteins are found on cell membranes and serve as the targets for V-SNARES (vesicle-SNAP receptors) permitting specific synaptic vesicle docking and fusion. A microdeletion in the region of chromosome 20 where this gene is located has been associated with pseudohypoparathyroidism type Ib. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

STX16
Polyclonal
Unconjugated
STX16
4930401D03; 5430410K23Rik; 6330500A18Rik; AI648908; AW553605; CG1467; CG1467-PA; Dm Syx16; Dmel\CG1467; Dmel_CG1467; DmSyx16; dSyntaxin16; dSyx 16; dSyx16; MGC90328; RP11-261P9.3; STX16; Syn16; syntaxin; syntaxin 16; Syntaxin16; syntaxin-16; Syx; SYX16; Syx16-PA
Sheep
Antigen affinity chromatography
RUO
8675
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
O14662
STX16
E. coli-derived recombinant human Syntaxin 16 isoform B, Leu165-Lys301.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.