missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STX12 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149230
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
SNARE that acts to regulate protein transport between late endosomes and the trans-Golgi network. The SNARE complex containing STX6, STX12, VAMP4 and VTI1A mediates vesicle fusion (in vitro).
Specifications
| STX12 | |
| Polyclonal | |
| Unconjugated | |
| STX12 | |
| AI850350; AU041521; MGC51957; Stx12; Stx13; STX14; Syn13; syntaxin 12; syntaxin 13; Syntaxin-12; Syntaxin-13 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 100226, 23673 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q86Y82, Q9ER00 | |
| STX12 | |
| E.coli-derived recombinant human Syntaxin 12, Met1-Gln200. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction