missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Stx-CLPesticides Column Kits

Product Code. p-3214813
Encompass_Preferred
Click to view available options
Diameter (Metric) Inner:
0.25 mm
This item is not returnable. View return policy

Product Code. 12945719

Brand: Restek™ 11190

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Stx-CLPesticides column, Stx-CLPesticides2 column, Universal Angled Y; Press-Tight connector and a Siltek-deactivated guard column

Requires

  • Concentration in Solvent: 0.100g/dL each in water
  • Volume: 1mL/ampule
  • Shelf Life: 6 months

Compounds

  • Acetaldehyde (75-07-0)
  • Acetone (67-64-1)
  • Acetonitrile (75-05-8)
  • Ethanol (64-17-5)
  • Ethyl acetate (141-78-6)
  • 2-Propanol (Isopropanol) (67-63-0)
  • Methanol (67-56-1)
  • Methyl ethyl ketone (78-93-3)

Application-specific columns for organochlorine pesticides and herbicides.

  • Baseline separations in less than 10 minutes
  • Siltek surface deactivation enhances responses for endrin, DDT, methoxychlor
  • Highly inert
  • Stable to 330°C

Note:
Columns are not preconnected in this kit.

Order Info

Custom lengths and film thicknesses available.

Specifications

Max. Temperature 330°C
Diameter (Metric) Inner 0.25 mm
Stationary Phase Stx™ -CLPesticides
For Use With (Application) Organochlorine Pesticides Analysis, Herbicides Analysis
Length (Metric) 30 m
Packing Material Stx™ -CLPesticides
Includes Rtx™-CLPesticides column, Rtx™-CLPesticides2 column, deactivated universal angled “Y” Press-Tight™ connector, guard column
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.