missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ STX 64

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
STX 64 is a potent steroid sulfatase inhibitor (IC50 = 8 nM). Estrone sulfate-stimulated uterine growth and mammary tumor growth are inhibited by STX 64 in animal models. STX 64 shows inhibition of steroid sulfatase activity in peripheral blood lymphocytes, liver and tumor tissues and is active in vivo. STX 64 is also a potent inhibitor of carbonic anhydrase II (IC50 = 25 nM).
Specifications
Specifications
| Chemical Name or Material | 6,7,8,9,10,11-Hexahydro-6-oxobenzo[b]cyclohepta[d]pyran-3-yl sulfamate |
| CAS | 288628-05-7 |
| Quantity | 25 mg |
| Target | Hydrolase Inhibitors |
| Molecular Formula | C14H15NO5S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction