Learn More
American Educational Products Student Project Planetarium
Ideal for individual study projects.
Brand:  American Educational Products 160
Product Code. 13169350
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Explains certain phenomena including day and night, season changes, the phases of the moon, and the locations of the planets.
  • The sun and Earth are mounted on the chart
  • Illustrated study guide includes planet position tables
  • Dimensions: 25.5 × 17 in. (64.7 × 42cm)