Learn More
Cole-Parmer™ Stuart™ Simultaneous Coagulant Doser
Click to view available options
For Use With (Equipment):
SW6 Flocculator
Unit Size:
Each
Description
- Promotes the clumping of fine floc into larger floc to allow easier separation from the water
Specifications
Specifications
| For Use With (Equipment) | SW6 Flocculator |