Learn More
Cole-Parmer™ Stuart™ Rotisserie for Hybridization Oven
Click to view available options
No. of Positions:
2
7
Unit Size:
Each
Description
- Choose from a wide range of accessories for your Cole-Parmer hybridization incubator.
- Select from differnt capacity rotisseries and bottles to suit your specific applicaiton.
Specifications
Specifications
| Product Type | Rotisserie |
| For Use With (Equipment) | Hybridization Bottles: 39454-98 |
| No. of Positions | 2 |