Learn More
Cole-Parmer™ Stuart™ Platform
Product Code. 11476608
Click to view available options
Dimensions (L x W):
340 x 300 mm
510 x 510 mm
Unit Size:
Each
Product Code. 11476608
Brand: Cole-Parmer™ 5170095
Only null left
Add to basket
Add to basket
Holds Erhlenmeyer Flasks or Bottles using rubber securing bars. Stuart™ Platforms with flexible cradle system allows different combinations of vessels to be used, offering maximum versatility. For use with Stuart™ Orbital Shakers (SSL1, SSM1)
  • For use with Stuart&;trade; Orbital Shakers (SSL1, SSM1)
  • Holds up Erhlenmeyer Flasks or Bottles using rubber securing bars.
  • Flexible cradle system has the benefit that different combinations of vessels can be used offering maximum versatility.
Specifications
Product Type Platform
Dimensions (L x W) 510 x 510 mm
For Use With (Application) For SGM-265-010C
For Use With (Equipment) Sutart™ Orbirtal Shakers SSM1, SSL1