Learn More
Cole-Parmer™ Stuart™ Platform
Product Code. 11456608
Click to view available options
Dimensions (L x W):
340 x 300 mm
510 x 510 mm
Unit Size:
Each
Product Code. 11456608
Brand: Cole-Parmer™ 5170098
Only null left
Add to basket
Add to basket
Holds Erhlenmeyer Flasks or Bottles using rubber securing bars. Stuart™ Platforms with flexible cradle system allows different combinations of vessels to be used, offering maximum versatility. For use with Stuart™ Orbital Shakers (SSL1, SSM1)
  • For use with Stuart™ Orbital Shakers (SSL1, SSM1)
  • Holds up Erhlenmeyer Flasks or Bottles using rubber securing bars.
  • Flexible cradle system has the benefit that different combinations of vessels can be used offering maximum versatility.
Specifications
Product Type Platform
Dimensions (L x W) 340 x 300 mm
For Use With (Application) For SGM-250-030K Large
For Use With (Equipment) Sutart™ Orbirtal Shakers SSM1, SSL1