missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Stuart™ Flat Lid

Product Code. 12717969
Click to view available options
For Use With (Equipment):
12L Bath
18 and 26L Bath
48L Bath
8L Bath
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12717969

Brand: Cole-Parmer™ 1212289

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Enhance the performance of your Techne baths and circulators

  • Lids, trays, temperature probes, and other accessories for your bath.

  • Manufactured from stainless steel and available to fit 8 L baths to help prevent evaporation losses.

Specifications

For Use With (Equipment) 12L Bath
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.