Learn More
Cole-Parmer™ Stuart™ Block Heater Insert
Click to view available options
No. of Places:
96
Unit Size:
Each
Description
- For use with Stuart block heaters
- Manufactured from anodized aluminum
- Separate hole accommodates a thermometer if desired
Specifications
Specifications
| Description | Holds 96-well plate |
| Holds | 96-well plate |
| Material | Anodized Aluminum |
| No. of Places | 96 |
| Type | Block |